Order Entry
United States
ContactUsLinkComponent
Nortriptyline Hydrochloride, LoGiCal, LGC Standards
Nortriptyline Hydrochloride, LoGiCal, LGC Standards
Catalog # 77697-054
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Nortriptyline Hydrochloride, LoGiCal, LGC Standards
Catalog # 77697-054
Supplier:  LGC Standards
Supplier Number:  LGCFOR0079.00
New Product
CAS Number:  

Specifications

  • CAS No.:
    894-71-3
  • Description:
    Nortriptyline Hydrochloride
  • Size:
    10 mg
  • Formula:
    C₁₉H₂₁N
  • Molecular Weight:
    299.84 g/mol
  • Cat. No.:
    77697-054

Specifications

About this item