Order Entry
United States
ContactUsLinkComponent
JWH-098 ((4-Methoxynaphthalen-1-yl)(2-methyl-1-pentylindol-3-yl)methanone) 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
JWH-098 ((4-Methoxynaphthalen-1-yl)(2-methyl-1-pentylindol-3-yl)methanone) 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-985
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
JWH-098 ((4-Methoxynaphthalen-1-yl)(2-methyl-1-pentylindol-3-yl)methanone) 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-985
Supplier:  LGC Standards
Supplier Number:  LGCAMP1275.73-11
New Product
CAS Number:  

Specifications

  • CAS No.:
    316189-74-9
  • Description:
    JWH-098 ((4-Methoxynaphthalen-1-yl)(2-methyl-1-pentylindol-3-yl)methanone) 1.0 mg/ml in Acetonitrile
  • Size:
    1 ml
  • Cat. No.:
    77696-985

Specifications

About this item