Ethylphenidate Hydrochloride 1.0 mg/ml in Acetonitrile (as free base), LoGiCal, LGC Standards
Catalog # 77696-956
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:19716-79-1
- Description:Ethylphenidate Hydrochloride 1.0 mg/ml in Acetonitrile (as free base)
- Size:1 ml
- Formula:C₁₅H₂₂ClNO₂
- Molecular Weight:283.8 g/mol
- Cat. No.:77696-956