Oxazepam-D5 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-855
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:65854-78-6
- Description:Oxazepam-D5 0.1 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₅H₁₁ClN₂O₂
- Cat. No.:77696-855