Methacrylic acid-methyl ester, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-361
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:80-62-6
- Description:Methacrylic acid-methyl ester
- Size:250 mg
- Formula:CH₂=C(CH₃)COOCH₃
- Molecular Weight:100.12 g/mol
- Cat. No.:77696-361