Fexofenadine hydrochloride 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-095
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:153439-40-8
- Description:Fexofenadine hydrochloride 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₃₂H₃₉NO₄
- Molecular Weight:538.13 g/mol
- Cat. No.:77696-095