Anti-GSC2 Rabbit Polyclonal Antibody
100 µg, Vial
Catalog # 77699-651
Supplier: AFG Bioscience
Supplier #: A14236-100UG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: GSC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Size:100 µg
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77699-651
- Reactivity:Human|Mouse
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice
- Antigen Symbol:GSC2
- Gene ID:O15499
- Immunogen:Synthesized peptide derived from the C-terminal region of Human GSC2.
- Conjugation:Unconjugated
- Antigen Synonyms:GSC2|GSCL|GSC-2|GSC-L|Homeobox protein goosecoid-2|Homeobox protein goosecoid-like
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



