Anti-GUCY1B1 Rabbit Polyclonal Antibody
77699-584
New Product
- Antigen Symbol:GUCY1B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q02153
- Antigen Synonyms:Soluble guanylate cyclase small subunit|Guanylate cyclase soluble subunit beta-1|GUCY1B1|GCS beta 1|GCS beta 3|GCS-beta-1|GCbeta1|GUCY1B3|beta 3|Gucy1b1|GC-SB3|Guanylate cyclase soluble beta 1 chain|Guanylate cyclase soluble subunit beta-3|SGC|GUCSB3|GC-S-beta-1|GCYB1_HUMAN|soluble|GCS-beta-3|GUC1B3|guanylate cyclase 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human GCS-β-1.
- Purification:Antigen affinity purified
- Cat. No.:77699-584
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: GUCY1B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat