Anti-S1PR2 Rabbit Polyclonal Antibody
Catalog # 77699-403
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:S1PR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O95136
- Antigen Synonyms:Sphingosine 1-phosphate receptor Edg-5|S1pr2|Endothelial differentiation G-protein coupled receptor 5|S1P receptor Edg 5|AGR16|Lysophospholipid receptor B2|H218|Endothelial differentiation sphingolipid G protein coupled receptor 5|Endothelial Cell Differentiation Gene 5|Sphingosine 1-phosphate receptor 2|S1P receptor 2|EDG 5|S1PR2_HUMAN|EDG5|Sphingosine 1 phosphate receptor Edg 5|Endothelial differentiation G protein coupled receptor 5|LPb2|S1P receptor EDG5|S1P receptor Edg-5|S1P2|Gpcr13
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human EDG-5.
- Purification:Antigen affinity purified
- Cat. No.:77699-403
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: S1PR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat