Anti-DAXX Rabbit Polyclonal Antibody
Catalog # 77699-327
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DAXX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9UER7
- Antigen Synonyms:Death domain associated protein 6|DAP 6|BING 2|Fas death domain-associated protein|MGC126246|BING2|Death associated protein 6|ETS1 associated protein 1|EAP1|Death domain associated protein|Fas death domain associated protein|DAXX_HUMAN|hDaxx|EAP 1|ETS1-associated protein 1|CENP-C binding protein|DAP6|MGC126245|Death domain-associated protein 6|Daxx|Fas binding protein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Daxx.
- Purification:Antigen affinity purified
- Cat. No.:77699-327
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: DAXX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat