Anti-CSTF1 Rabbit Polyclonal Antibody
Catalog # 77699-248
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CSTF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q05048
- Antigen Synonyms:Cleavage stimulation factor|50kD|50kDa|cleavage stimulation factor|subunit 1|50 kD|CSTF 50 kDa subunit|CstFp50|CSTF1_HUMAN|Cleavage stimulation factor 50 kDa subunit|3 prime pre RNA|CstF 50|CF-1 50 kDa subunit|OTTHUMP00000031332|CstF-50|Cleavage stimulation factor 50kDa subunit|CSTF1|CF 1 50 kDa subunit|3' pre RNA|Cleavage stimulation factor subunit 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human CstF-50.
- Purification:Antigen affinity purified
- Cat. No.:77699-248
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: CSTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat