Anti-CRHR1 Rabbit Polyclonal Antibody
77699-232
New Product
- Antigen Symbol:CRHR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P34998
- Antigen Synonyms:CRH-R1|crf receptor 1|CRH-R-1|CRF R|CRF1|CRFR|CRF-R|CRHR|crf type 1|Crhr1|Corticotropin-releasing hormone receptor 1|Corticotropin releasing factor type 1 receptor|CRF-R1|CRFR1|CRHR1L|crf receptor type 1|CRH-R1h|Corticotropin-releasing factor receptor 1|CRFR-1|corticotropin releasing hormone receptor 1|CRF-R-1|CRHR1f|CRFR1_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CRF-RI.
- Purification:Antigen affinity purified
- Cat. No.:77699-232
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:5000
Type:
Antigen: CRHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat