Anti-CDC6 Rabbit Polyclonal Antibody
Catalog # 77699-122
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CDC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q99741
- Antigen Synonyms:S. cerevisiae|Cdc 18L|Cdc18L|homolog) like|CDC6_HUMAN|HsCDC18|Cell division control protein 6|Cdc18-related protein|p62(cdc6)|Cdc18 related protein|HsCDC6|CDC6-related protein|Cdc 6|CDC6 cell division cycle 6 homolog|CELL CYCLE CONTROLLER CDC6|Cell division cycle 6 homolog|Cdc6p|Cell division cycle 6|CDC18(S.pombe)|p62|CDC6 related protein|Cell division cycle 6 homolog (S. cerevisiae)|CDC18 (S.pombe)|CDC18 (cell division cycle 18|homolog of|cdc6|p62(cdc 6)|HsCDC 6|S.pombe|Cell division control protein 6 homolog
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Cdc6 around the non-phosphorylation site of S54.
- Purification:Antigen affinity purified
- Cat. No.:77699-122
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: CDC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse