Anti-CDC34 Rabbit Polyclonal Antibody
Catalog # 77699-117
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CDC34
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P49427
- Antigen Synonyms:Ubiquitin-protein ligase R1|Cdc34|UBE2 R1|E2 CDC34|Cell division cycle 34|Ubiquitin carrier protein|Ubiquitin conjugating enzyme Cdc34|UBC3|UBC 3|Ubiquitin protein ligase|Ubiquitin conjugating enzyme E2 32 kDa complementing|Ubiquitin protein ligase R1|UBCH3|Cdc 34|Cell division cycle 34 homolog (S. cerevisiae)|Ubiquitin-conjugating enzyme E2-CDC34|Ubiquitin-conjugating enzyme E2-32 kDa complementing|Ubiquitin-conjugating enzyme E2 R1|UBE2R1|Cell division cycle 34 homolog|UB2R1_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Cdc34.
- Purification:Antigen affinity purified
- Cat. No.:77699-117
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: CDC34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat