Anti-ACKR4 Rabbit Polyclonal Antibody
Catalog # 77699-082
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACKR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9NPB9
- Antigen Synonyms:CCBP2|Orphan seven transmembrane receptor chemokine related|VSHK1|CCR10|Chemokine (C C motif) receptor like 1|ACKR4|CCRL1|C C chemokine receptor type 11|C-C chemokine receptor type 11|CCX CKR|CC chemokine receptor-like 1|CC chemokine receptor like 1|CC-CKR-11|CKRB|Chemokine cc motif receptor like protein 1|CC CKR 11|Chemokine receptor like 1|C-C CKR-11|CCR 11|CCCKR11|CCRL1_HUMAN|Chemocentryx chemokine receptor|PPR1|Atypical chemokine receptor 4|CCR-11|CKR11
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human CCRL1.
- Purification:Antigen affinity purified
- Cat. No.:77699-082
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:40000
Type:
Antigen: ACKR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse