Anti-C1QBP Rabbit Polyclonal Antibody
Catalog # 77699-000
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:C1QBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q07021
- Antigen Synonyms:GC1QBP|p32|Mitochondrial matrix protein p32|HABP1|C1q|Hyaluronan-binding protein 1|Pre mrna splicing factor SF2 P32 subunit precursor|ASF/SF2 associated protein p32|globular domain of|Hyaluronan binding protein 1|HABP 1|p32 splicing factor|Complement component 1 q subcomponent binding protein|C1QBP_HUMAN|C1qBP|GC1QR|mitochondrial|GC1q R protein|Complement component 1 Q subcomponent-binding protein|GC1Q R|GC1q-R protein|C1q globular domain binding protein|p33|Complement component 1 Q subcomponent binding protein mitochondrial|receptor for|SF2p32|Splicing factor SF2 associated protein|Glycoprotein gC1qBP
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human C1QBP.
- Purification:Antigen affinity purified
- Cat. No.:77699-000
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: C1QBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat