Anti-MPC1 Rabbit Polyclonal Antibody
Catalog # 77698-985
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MPC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9Y5U8
- Antigen Synonyms:Mpc1|CGI129|pyruvate carrier, Mitochondrial, 1|HSPC040 protein|PNAS 115|MPYCD|Apoptosis regulating basic protein|MPC1_HUMAN|Brain protein 44-like protein|dJ68L15.3|PNAS115|CGI 129|Brain protein 44 like protein|HSPC040|Mitochondrial pyruvate carrier 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human Brp44L.
- Purification:Antigen affinity purified
- Cat. No.:77698-985
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: MPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat