Anti-FGFR2 Rabbit Polyclonal Antibody
77698-959
New Product
- Antigen Symbol:FGFR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P21802
- Antigen Synonyms:TK14|CD332 antigen|receptor like 14|ECT1|Jackson Weiss syndrome|TK25|BEK|soluble FGFR4 variant 4|FGF receptor|K-sam|Keratinocyte growth factor receptor|JWS|KGFR|FGFR2_HUMAN|CFD1|BEK fibroblast growth factor receptor|BFR1|FGFR-2|FGFR 2|Fgfr2|Fibroblast growth factor receptor 2|Hydroxyaryl protein kinase|Keratinocyte growth factor receptor 2|protein tyrosine kinase|K SAM|CEK3|CD332|bacteria-expressed kinase|Craniofacial dysostosis 1|KSAM|BBDS
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Bek.
- Purification:Antigen affinity purified
- Cat. No.:77698-959
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: FGFR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat