Anti-NR2F2 Rabbit Polyclonal Antibody
77698-899
New Product
- Antigen Symbol:NR2F2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P24468
- Antigen Synonyms:COUP-TF II|Nuclear receptor subfamily 2 group F member 2|Apolipoprotein A-I regulatory protein 1|COUP-TF2|COT2_HUMAN|Apolipoprotein AI regulatory protein 1|ARP1|COUP transcription factor 2|COUP transcription factor II|ARP-1|COUP TF2|NR2F2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human ARP-1.
- Purification:Antigen affinity purified
- Cat. No.:77698-899
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: NR2F2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat