Anti-CHRNA5 Rabbit Polyclonal Antibody
Catalog # 77698-790
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CHRNA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P30532
- Antigen Synonyms:Cholinergic receptor neuronal nicotinic alpha polypeptide 5|Neuronal nicotinic acetylcholine receptor alpha 5 subunit|NACHRA 5|CHRNA 5|Neuronal nicotinic acetylcholine receptor, alpha5 subunit|Acetylcholine receptor nicotinic alpha 5 (neuronal)|Neuronal acetylcholine receptor subunit alpha-5|ACHA5_HUMAN|LNCR2|OTTHUMP00000185019|Neuronal acetylcholine receptor subunit alpha 5|AChR|Cholinergic receptor nicotinic alpha polypeptide 5|CHRNA5|Cholinergic receptor nicotinic alpha 5|NACHRA5
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human AChRα5.
- Purification:Antigen affinity purified
- Cat. No.:77698-790
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: CHRNA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey