Anti-TUBA1C Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100
Type:
Antigen: TUBA1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77711-035
- 77711-034
- Antigen Symbol:
- TUBA1C
- TUBA1C
- Antigen Synonyms:
- Tuba1c|M[a]6|tubulin, alpha 1c|TUBA6|Tubulin alpha-6 chain|Tubulin alpha 1C chain|Alpha tubulin 6|Tubulin alpha 6 chain|Alpha tubulin isotype M alpha 6|Alpha-tubulin 6|Tubulin alpha-1C chain|TBA1C_HUMAN|bcm948|tubulin alpha 6
- Tuba1c|M[a]6|tubulin, alpha 1c|TUBA6|Tubulin alpha-6 chain|Tubulin alpha 1C chain|Alpha tubulin 6|Tubulin alpha 6 chain|Alpha tubulin isotype M alpha 6|Alpha-tubulin 6|Tubulin alpha-1C chain|TBA1C_HUMAN|bcm948|tubulin alpha 6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BQE3
- Q9BQE3
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TUBA1C
- Fusion protein of Human TUBA1C
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together