Anti-STRADB Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:5000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: STRADB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77711-407
- 77711-408
- Antigen Symbol:
- STRADB
- STRADB
- Antigen Synonyms:
- ILP interacting protein ILPIPA|CALS-21|Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein|CALS 21|PRO1038|ILPIPA|STE20-related kinase adapter protein beta|ILP-interacting protein|STRAD beta|ILPIP|STRADB|STRAB_HUMAN|ALS2CR2|Amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 2|ALS2 chromosome region gene 2|PAPK|Likely ortholog of mouse polyploidy associated protein kinase|Pseudokinase ALS2CR2
- ILP interacting protein ILPIPA|CALS-21|Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein|CALS 21|PRO1038|ILPIPA|STE20-related kinase adapter protein beta|ILP-interacting protein|STRAD beta|ILPIP|STRADB|STRAB_HUMAN|ALS2CR2|Amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 2|ALS2 chromosome region gene 2|PAPK|Likely ortholog of mouse polyploidy associated protein kinase|Pseudokinase ALS2CR2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9C0K7
- Q9C0K7
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human STRADB
- Fusion protein of Human STRADB
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together