Anti-STRA8 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: STRA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77712-396
- 77712-397
- Antigen Symbol:
- STRA8
- STRA8
- Antigen Synonyms:
- Stimulated by retinoic acid Gene 8|stimulated by retinoic acid gene 8 homolog|STRA 8|STRA8_HUMAN|Stimulated by retinoic acid 8|Stimulated by retinoic acid gene 8 protein homolog|STRA8|stimulated by retinoic acid 8 homolog
- Stimulated by retinoic acid Gene 8|stimulated by retinoic acid gene 8 homolog|STRA 8|STRA8_HUMAN|Stimulated by retinoic acid 8|Stimulated by retinoic acid gene 8 protein homolog|STRA8|stimulated by retinoic acid 8 homolog
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z7C7
- Q7Z7C7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human STRA8
- Synthetic peptide of Human STRA8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together