Anti-STMN1 Rabbit Polyclonal Antibody
Catalog # 77703-019
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:STMN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P16949
- Antigen Synonyms:Pr22 protein|Op18|Protein Pr22|Oncoprotein 18|SMN|Stathmin1|C1orf215|Metablastin|OP 18|LAP18|Stmn1|Prosolin|STMN 1|p19|Stathmin|LAP 18|Lag|pp19|PR22|Leukemia-associated phosphoprotein p18|Phosphoprotein 19|STMN1_HUMAN|pp17|p18|Leukemia associated phosphoprotein p18|Phosphoprotein p19
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Op18 around the non-phosphorylation site of S62.
- Purification:Antigen affinity purified
- Cat. No.:77703-019
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: STMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat