Anti-SOX2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77689-219
- 77711-165
- Antigen Symbol:
- SOX2
- SOX2
- Antigen Synonyms:
- SRY box containing gene 2|Transcription factor SOX 2|Sox2|MCOPS3|MGC2413|ANOP3|cb236|SRY related HMG box gene 2|RGD1565646|SRY (sex determining region Y) box 2|Sex determining region Y box 2|SRY-box 2|ysb|SRY related HMG box 2|Transcription factor SOX-2|SOX 2|Delta EF2a|SOX2_HUMAN|lcc|MGC148683
- SRY box containing gene 2|Transcription factor SOX 2|Sox2|MCOPS3|MGC2413|ANOP3|cb236|SRY related HMG box gene 2|RGD1565646|SRY (sex determining region Y) box 2|Sex determining region Y box 2|SRY-box 2|ysb|SRY related HMG box 2|Transcription factor SOX-2|SOX 2|Delta EF2a|SOX2_HUMAN|lcc|MGC148683
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48431
- P48431
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Transcription factor SOX-2 protein (1-317AA)
- Recombinant Human Transcription factor SOX-2 protein (1-317AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together