Anti-SLC39A1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC39A1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77712-050
- 77712-049
- Antigen Symbol:
- SLC39A1
- SLC39A1
- Antigen Synonyms:
- ZIP-1|IRT1|ZIP1|SLC39A1|Zinc transporter ZIP1|Zinc-iron-regulated transporter-like|ZIRTL|Zrt- and Irt-like protein 1|S39A1_HUMAN|hZIP1|Solute carrier family 39 member 1
- ZIP-1|IRT1|ZIP1|SLC39A1|Zinc transporter ZIP1|Zinc-iron-regulated transporter-like|ZIRTL|Zrt- and Irt-like protein 1|S39A1_HUMAN|hZIP1|Solute carrier family 39 member 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NY26
- Q9NY26
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Zinc transporter ZIP1 protein (126-179AA)
- Recombinant Human Zinc transporter ZIP1 protein (126-179AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together