Anti-SEMA4A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: SEMA4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77711-116
- 77711-117
- Antigen Symbol:
- SEMA4A
- SEMA4A
- Antigen Synonyms:
- SEMAB|Semaphorin 4A precursor|CORD10|Sema domain immunoglobulin domain Ig transmembrane domain TM and short cytoplasmic domain semaphorin 4A|SEMB|Semaphorin B|Semaphorin-B|Sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin)|Semaphorin-4A|RP11 54H19 2|SEM4A_HUMAN|Sema domain immunoglobulin domain Ig transmembrane domain TM and short cytoplasmic domain 4A|Sema B|RP35|Sema4a
- SEMAB|Semaphorin 4A precursor|CORD10|Sema domain immunoglobulin domain Ig transmembrane domain TM and short cytoplasmic domain semaphorin 4A|SEMB|Semaphorin B|Semaphorin-B|Sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin)|Semaphorin-4A|RP11 54H19 2|SEM4A_HUMAN|Sema domain immunoglobulin domain Ig transmembrane domain TM and short cytoplasmic domain 4A|Sema B|RP35|Sema4a
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H3S1
- Q9H3S1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human SEMA4A
- Fusion protein of Human SEMA4A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together