Anti-RHOB Rabbit Polyclonal Antibody
77702-973
New Product
- Antigen Symbol:RHOB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P62745
- Antigen Synonyms:RHOB_HUMAN|Rho related GTP binding protein RhoB Precursor|Rho cDNA clone 6|H6|Rhob|Rho-related GTP-binding protein RhoB|ARH6|ras homolog family member B|Aplysia RAS-related homolog 6|MSTP081|RHOH6|ARHB|ras homolog gene family|member B|oncogene RHO H6|MST081
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Rho B.
- Purification:Antigen affinity purified
- Cat. No.:77702-973
- Packaging:Vial
Type:
Antigen: RHOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat