Anti-rbsK Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: rbsK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
- Catalog No:
- 77689-246
- 77712-480
- Antigen Symbol:
- rbsK
- rbsK
- Antigen Synonyms:
- JW3731|Ribokinase|EC 2.7.1.15|RK|b3752|rbsK
- JW3731|Ribokinase|EC 2.7.1.15|RK|b3752|rbsK
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- E. coli
- E. coli
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0A9J6
- P0A9J6
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Escherichia coli Ribokinase protein (1-309AA)
- Recombinant Escherichia coli Ribokinase protein (1-309AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together