Anti-RBBP5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: RBBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77712-425
- 77712-426
- Antigen Symbol:
- RBBP5
- RBBP5
- Antigen Synonyms:
- Set1c WD40 repeat protein|Retinoblastoma binding protein RBQ3|homolog|RBBP 5|RBQ 3|Retinoblastoma-binding protein RBQ-3|RBBP5_HUMAN|Retinoblastoma binding protein5|RBBP5|Retinoblastoma-binding protein 5|Retinoblastoma binding protein 5|SWD1|RBBP-5|RBQ3
- Set1c WD40 repeat protein|Retinoblastoma binding protein RBQ3|homolog|RBBP 5|RBQ 3|Retinoblastoma-binding protein RBQ-3|RBBP5_HUMAN|Retinoblastoma binding protein5|RBBP5|Retinoblastoma-binding protein 5|Retinoblastoma binding protein 5|SWD1|RBBP-5|RBQ3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15291
- Q15291
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human RBBP5
- Fusion protein of Human RBBP5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together