Anti-PRKCH Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: PRKCH
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77710-761
- 77710-762
- Antigen Symbol:
- PRKCH
- PRKCH
- Antigen Synonyms:
- nPKC-eta|PKC-L|PKCL|MGC26269|PKC L|MGC5363|KPCL_HUMAN|PKCh|MGC 5363|Protein kinase C eta type|PRKCL|PKC h|Prkch|nPKC eta|Protein kinase C eta
- nPKC-eta|PKC-L|PKCL|MGC26269|PKC L|MGC5363|KPCL_HUMAN|PKCh|MGC 5363|Protein kinase C eta type|PRKCL|PKC h|Prkch|nPKC eta|Protein kinase C eta
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P24723
- P24723
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Protein kinase C eta type protein (1-678AA)
- Recombinant Human Protein kinase C eta type protein (1-678AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together