Anti-OR4N4 Rabbit Polyclonal Antibody
Catalog # 77702-640
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:OR4N4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q8N0Y3
- Antigen Synonyms:Olfactory receptor OR15-5|Olfactory receptor OR15-1|OR4N4|Olfactory receptor 4N4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Olfactory receptor 4N4.
- Purification:Antigen affinity purified
- Cat. No.:77702-640
- Packaging:Vial
Specifications
About this item
Type:
Antigen: OR4N4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human