Anti-OR2AK2 Rabbit Polyclonal Antibody
Catalog # 77703-009
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:OR2AK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q8NG84
- Antigen Synonyms:Olfactory receptor OR1-47|OR2AK1P|Olfactory receptor 2AK1|Olfactory receptor 2AK2|OR2AK2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human Olfactory receptor 2AK2.
- Purification:Antigen affinity purified
- Cat. No.:77703-009
- Packaging:Vial
Specifications
About this item
Type:
Antigen: OR2AK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human