Anti-NEU1 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NEU1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77711-583
- 77711-584
- Antigen Symbol:
- NEU1
- NEU1
- Antigen Synonyms:
- NEU|Sialidase|Neuraminidase|lysosomal|N-acetyl-alpha-neuraminidase 1|exo-alpha-sialidase|SIAL1|Sialidase-1|G9 sialidase|sialidase 1 (lysosomal sialidase)|NEUR1_HUMAN|Sialidase 1|N acetyl alpha neuraminidase 1|NEU1|Acetylneuraminyl hydrolase|Lysosomal sialidase|NANH|Neuraminidase 1
- NEU|Sialidase|Neuraminidase|lysosomal|N-acetyl-alpha-neuraminidase 1|exo-alpha-sialidase|SIAL1|Sialidase-1|G9 sialidase|sialidase 1 (lysosomal sialidase)|NEUR1_HUMAN|Sialidase 1|N acetyl alpha neuraminidase 1|NEU1|Acetylneuraminyl hydrolase|Lysosomal sialidase|NANH|Neuraminidase 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99519
- Q99519
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sialidase-1 protein (201-415AA)
- Recombinant Human Sialidase-1 protein (201-415AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together