Anti-MAP3K9/MAP3K10 Rabbit Polyclonal Antibody
100 µg, Vial
Catalog # 77703-174
Supplier: AFG Bioscience
Supplier #: A24461-100UG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: MAP3K9/MAP3K10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Size:100 µg
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77703-174
- Reactivity:Human|Mouse
- Shipping Temperature:Shipped on blue ice
- Antigen Symbol:MAP3K9/MAP3K10
- Gene ID:P80192/Q02779
- Immunogen:Synthesized peptide derived from Human MLK1/2 around the non-phosphorylation site of T312/266.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:Protein kinase MST|MAP3K10|MLK2|MAP3K9|Mitogen-activated protein kinase kinase kinase 9|Mixed lineage kinase 1|MST|PRKE1|Mixed lineage kinase 2|MLK1|Mitogen-activated protein kinase kinase kinase 10
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



