Anti-KLRC1 Rabbit Polyclonal Antibody
Catalog # 77702-960
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KLRC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P26715
- Antigen Synonyms:Natural killer cell lectin|NKG2 1/B activating NK receptor|NKG2 B|NKG2 A|CD159a antigen|MGC13374|rNKG2A|Killer cell lectin like receptor subfamily C member 1|CD159 antigen-like family member A|NKG2B|Natural killer group protein 2|Natural killer cell protein group 2-A1|NK cell receptor A|C lectin type II protein|Natural killer cell receptor NKG2A|NKG2-1/B activating NK receptor|KLRC1|NKG2 A/B type II integral membrane protein|NKG2A_HUMAN|NKG2-A/B-activating NK receptor|CD159 antigen like family member A|NKG2|CD159a|NKG2 A/NKG2 B type II integral membrane protein|NKG2A|NKG2 A/B activating NK receptor|MGC59791|NKG2-A/NKG2-B type II integral membrane protein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human CD159a.
- Purification:Antigen affinity purified
- Cat. No.:77702-960
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: KLRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human