Anti-KLK13 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, IHC:1:25-1:100
Type:
Antigen: KLK13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77712-431
- 77712-432
- Antigen Symbol:
- KLK13
- KLK13
- Antigen Synonyms:
- Kallikrein like gene 4|KLKL 4|Kallikrein 13|Kallikrein like 4 variant included|KLK13_HUMAN|Kallikrein related peptidase 13|Kallikrein like protein 4|Kalikrein|Kallikrein-13|KLK-L4|Kallikrein-like protein 4|KLK13|Kallikrein Like 4|Kallikrein L4|KLKL4|DKFZp586J1923
- Kallikrein like gene 4|KLKL 4|Kallikrein 13|Kallikrein like 4 variant included|KLK13_HUMAN|Kallikrein related peptidase 13|Kallikrein like protein 4|Kalikrein|Kallikrein-13|KLK-L4|Kallikrein-like protein 4|KLK13|Kallikrein Like 4|Kallikrein L4|KLKL4|DKFZp586J1923
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKR3
- Q9UKR3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human KLK13
- Fusion protein of Human KLK13
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together