Anti-KLF15 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: KLF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77711-220
- 77711-219
- Antigen Symbol:
- KLF15
- KLF15
- Antigen Synonyms:
- Krueppel-like factor 15|KLF15_HUMAN|KLF15|Kruppel like factor 15|KKLF protein|KLF 15|Kidney enriched Kruppel like factor|Kidney-enriched krueppel-like factor|KKLF
- Krueppel-like factor 15|KLF15_HUMAN|KLF15|Kruppel like factor 15|KKLF protein|KLF 15|Kidney enriched Kruppel like factor|Kidney-enriched krueppel-like factor|KKLF
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UIH9
- Q9UIH9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human KLF15
- Fusion protein of Human KLF15
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together