Anti-KCNK17 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:100-1:300
Type:
Antigen: KCNK17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77712-351
- 77712-352
- Antigen Symbol:
- KCNK17
- KCNK17
- Antigen Synonyms:
- TWIK related acid sensitive K(+) channel 4|TALK 2|KCNKH_HUMAN|KCNK17|acid sensitive potassium channel protein TASK 4|TASK 4|K2p17.1|2P domain potassium channel Talk-2|TALK2|TWIK related alkaline pH activated K(+) channel 2|Potassium channel subfamily K member 17|Potassium channel, subfamily K, member 17|TALK-2|TWIK-related acid-sensitive K(+) channel 4|TWIK-related alkaline pH-activated K(+) channel 2|2P domain potassium channel Talk 2|Acid-sensitive potassium channel protein TASK-4|TASK4
- TWIK related acid sensitive K(+) channel 4|TALK 2|KCNKH_HUMAN|KCNK17|acid sensitive potassium channel protein TASK 4|TASK 4|K2p17.1|2P domain potassium channel Talk-2|TALK2|TWIK related alkaline pH activated K(+) channel 2|Potassium channel subfamily K member 17|Potassium channel, subfamily K, member 17|TALK-2|TWIK-related acid-sensitive K(+) channel 4|TWIK-related alkaline pH-activated K(+) channel 2|2P domain potassium channel Talk 2|Acid-sensitive potassium channel protein TASK-4|TASK4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96T54
- Q96T54
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human KCNK17
- Fusion protein of Human KCNK17
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together