Anti-GRK4 Rabbit Polyclonal Antibody
Catalog # 77702-675
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GRK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:P32298
- Antigen Synonyms:GPRK4|G protein-coupled receptor kinase 2 (Drosophila)-like|GRK4|ITI1|G protein-coupled receptor kinase 4|GRK4_HUMAN|G protein coupled receptor kinase 4|GPRK2L|GRK4a|G protein-coupled receptor kinase GRK4|G proteinm coupled receptor kinase GRK4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human GRK 4.
- Purification:Antigen affinity purified
- Cat. No.:77702-675
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: GRK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human