Anti-GRIN2C Rabbit Polyclonal Antibody
Catalog # 77702-712
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:GRIN2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q14957
- Antigen Synonyms:Glutamate Receptor Ionotropic N Methyl D Aspartate 2C|NMDAR2C|N Methyl D Aspartate Receptor Subtype 2C|N Methly D Aspartate Receptor Channel Subunit Epsilon 3|Grin2c|N-methyl D-aspartate receptor subtype 2C|N methyl D aspartate receptor subunit 2C|GluN2C|Glutamate receptor, ionotropic, N methyl D aspartate 2C|Glutamate receptor ionotropic, NMDA 2C|NMDE3_HUMAN|NR2C|OTTHUMP00000041930|Glutamate [NMDA] receptor subunit epsilon-3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human NMDAε3.
- Purification:Antigen affinity purified
- Cat. No.:77702-712
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: GRIN2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human