Anti-FGF9 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200
Type:
Antigen: FGF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77711-530
- 77711-531
- Antigen Symbol:
- FGF9
- FGF9
- Antigen Synonyms:
- FGF 9|Glia-activating factor|MGC119914|HBFG 9|FGF-9|FGF9_HUMAN|FGF9|GAF (Glia-activafibroblast growth factor 9 (glia-activating factor)|GAF|HBFG9|Heparin-binding growth factor 9|Glia Activating Factor|HBGF-9|Fibroblast growth factor 9|MGC119915|SYNS3
- FGF 9|Glia-activating factor|MGC119914|HBFG 9|FGF-9|FGF9_HUMAN|FGF9|GAF (Glia-activafibroblast growth factor 9 (glia-activating factor)|GAF|HBFG9|Heparin-binding growth factor 9|Glia Activating Factor|HBGF-9|Fibroblast growth factor 9|MGC119915|SYNS3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31371
- P31371
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human FGF9
- Fusion protein of Human FGF9
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together