Anti-FASTKD1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:10000, IHC:1:100-1:300
Type:
Antigen: FASTKD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77712-278
- 77712-277
- Antigen Symbol:
- FASTKD1
- FASTKD1
- Antigen Synonyms:
- FLJ21901|OTTHUMP00000207010|KIAA1800|FASTKD1|OTTHUMP00000207008|FAKD1_HUMAN|FAST kinase domain-containing protein 1|FAST kinase domains 1|FAST kinase domain containing protein 1
- FLJ21901|OTTHUMP00000207010|KIAA1800|FASTKD1|OTTHUMP00000207008|FAKD1_HUMAN|FAST kinase domain-containing protein 1|FAST kinase domains 1|FAST kinase domain containing protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53R41
- Q53R41
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human FASTKD1
- Fusion protein of Human FASTKD1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together