Anti-DUSP13 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: DUSP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77711-849
- 77711-850
- Antigen Symbol:
- DUSP13
- DUSP13
- Antigen Synonyms:
- DUSP13|Dual specificity phosphatase SKRP4|DUSP13B|TMDPDual specificity protein phosphatase 13 isoform B|EC 3.1.3.16|EC 3.1.3.48|Testis- and skeletal-muscle-specific DSP
- DUSP13|Dual specificity phosphatase SKRP4|DUSP13B|TMDPDual specificity protein phosphatase 13 isoform B|EC 3.1.3.16|EC 3.1.3.48|Testis- and skeletal-muscle-specific DSP
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UII6
- Q9UII6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human DUSP13
- Fusion protein of Human DUSP13
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together