Anti-DNAJA1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200
Type:
Antigen: DNAJA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77712-140
- 77712-141
- Antigen Symbol:
- DNAJA1
- DNAJA1
- Antigen Synonyms:
- OTTHUMP00000021193|subfamily A|HDJ-2|DJ 2|DNAJ2|Heat shock 40 kDa protein 4|heat shock protein DNAJ like 2|HSJ-2|DnaJ (Hsp40) homolog|Heat shock protein J2|HSPF4|DNJA1_HUMAN|DjA1|HDJ2|Neural precursor cell expressed developmentally down regulated 7|DJ2|HSDJ|member 1|NEDD7|HSJ2|hDJ 2|DnaJ protein homolog 2|Human DnaJ protein 2|DnaJ homolog subfamily A member 1|Dnaja1
- OTTHUMP00000021193|subfamily A|HDJ-2|DJ 2|DNAJ2|Heat shock 40 kDa protein 4|heat shock protein DNAJ like 2|HSJ-2|DnaJ (Hsp40) homolog|Heat shock protein J2|HSPF4|DNJA1_HUMAN|DjA1|HDJ2|Neural precursor cell expressed developmentally down regulated 7|DJ2|HSDJ|member 1|NEDD7|HSJ2|hDJ 2|DnaJ protein homolog 2|Human DnaJ protein 2|DnaJ homolog subfamily A member 1|Dnaja1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31689
- P31689
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human DNAJA1
- Fusion protein of Human DNAJA1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together