Anti-CYP3A4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:50-1:200
Type:
Antigen: CYP3A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77711-972
- 77711-971
- Antigen Symbol:
- CYP3A4
- CYP3A4
- Antigen Synonyms:
- P450C3|NF25|cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3|1,8-cineole 2-exo-monooxygenase|Quinine 3 monooxygenase|Quinine 3-monooxygenase|Cytochrome P450 HLp|Taurochenodeoxycholate 6 alpha hydroxylase|Taurochenodeoxycholate 6-alpha-hydroxylase|Cytochrome P450 subfamily IIIA polypeptide 4|cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4|CP34|Cytochrome P450-PCN1|CYP3|Nifedipine oxidase|MGC126680|P450, family III|CYP3A|Albendazole sulfoxidase|Cytochrome P450 3A4|NF 25|Albendazole monooxygenase|CYP3A4|Cytochrome P450 3A3|CYPIIIA4|CYP3A3|CP3A4_HUMAN|CYPIIIA3|P450 III steroid inducible|Cytochrome P450 NF-25|P450 PCN1|HLP|Glucocorticoid inducible P450|P450PCN1|Cytochrome P450 family 3 subfamily A polypeptide 4|CP33
- P450C3|NF25|cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3|1,8-cineole 2-exo-monooxygenase|Quinine 3 monooxygenase|Quinine 3-monooxygenase|Cytochrome P450 HLp|Taurochenodeoxycholate 6 alpha hydroxylase|Taurochenodeoxycholate 6-alpha-hydroxylase|Cytochrome P450 subfamily IIIA polypeptide 4|cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4|CP34|Cytochrome P450-PCN1|CYP3|Nifedipine oxidase|MGC126680|P450, family III|CYP3A|Albendazole sulfoxidase|Cytochrome P450 3A4|NF 25|Albendazole monooxygenase|CYP3A4|Cytochrome P450 3A3|CYPIIIA4|CYP3A3|CP3A4_HUMAN|CYPIIIA3|P450 III steroid inducible|Cytochrome P450 NF-25|P450 PCN1|HLP|Glucocorticoid inducible P450|P450PCN1|Cytochrome P450 family 3 subfamily A polypeptide 4|CP33
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08684
- P08684
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CYP3A4
- Fusion protein of Human CYP3A4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together