Anti-Cleaved-LMNA (N231) Rabbit Polyclonal Antibody
Catalog # 77703-052
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Cleaved-LMNA (N231)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P02545
- Antigen Synonyms:LMNL1|LMNA|FPLD|FPL|IDC|PRO1|LMNC|CMT2B1|LGMD1B|LMN1|CDCD1|70 kDa lamin|Lamin A/C like 1|LMN 1|Lamin-A/C|LFP|FPLD2|Lamin A/C|LMNA_HUMAN|LDP1|Renal carcinoma antigen NY REN 32|Renal carcinoma antigen NY-REN-32|Prelamin A/C|lamin-a|Lamin A|CDDC|LMN C|CMD1A|EMD2|Cardiomyopathy dilated 1A (autosomal dominant)|Lamin C|Renal carcinoma antigen NYREN32|Lamin|HGPS|LMN A|Limb girdle muscular dystrophy 1B (autosomal dominant)
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Lamin A.
- Purification:Antigen affinity purified
- Cat. No.:77703-052
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: Cleaved-LMNA (N231)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat