Anti-CAPN1 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:5000, WB:1:200-1:1000
Type:
Antigen: CAPN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77712-248
- 77712-249
- Antigen Symbol:
- CAPN1
- CAPN1
- Antigen Synonyms:
- CANPL 1|large polypeptide L1|Calcium activated neutral proteinase small subunit|CAPN 1|muCANP|CAPN1|CANP 1|CANP small subunit|Micromolar Calpain|Calcium dependent protease small subunit 1|Micromolar-calpain|Calpain mu-type|Calpain-1 large subunit|Cell proliferation inducing protein 30|Calcium dependent protease small subunit|Mu Calpain|Calpain mu type|CANP|Calpain small subunit 1|Ca2 activated neutral protease|Calpain 1 large subunit|CANPL1|Calpain 1|Calpain regulatory subunit|Calpain|Calpain-1 catalytic subunit|CAN1_HUMAN|(mu/I) large subunit|CANP1|Calcium-activated neutral proteinase 1|Calcium activated neutral proteinase|muCL|Cell proliferation-inducing gene 30 protein
- CANPL 1|large polypeptide L1|Calcium activated neutral proteinase small subunit|CAPN 1|muCANP|CAPN1|CANP 1|CANP small subunit|Micromolar Calpain|Calcium dependent protease small subunit 1|Micromolar-calpain|Calpain mu-type|Calpain-1 large subunit|Cell proliferation inducing protein 30|Calcium dependent protease small subunit|Mu Calpain|Calpain mu type|CANP|Calpain small subunit 1|Ca2 activated neutral protease|Calpain 1 large subunit|CANPL1|Calpain 1|Calpain regulatory subunit|Calpain|Calpain-1 catalytic subunit|CAN1_HUMAN|(mu/I) large subunit|CANP1|Calcium-activated neutral proteinase 1|Calcium activated neutral proteinase|muCL|Cell proliferation-inducing gene 30 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P07384
- P07384
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CAPN1
- Fusion protein of Human CAPN1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together