Anti-BCL6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: BCL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77711-547
- 77711-546
- Antigen Symbol:
- BCL6
- BCL6
- Antigen Synonyms:
- B cell lymphoma 6 protein|ZBTB 27|Bcl 6|BCL 5|BCL6|BCL5|Zinc finger and BTB domain containing protein 27|Lymphoma Associated Zinc Finger Gene On Chromosome 3 (LAZ3)|Zinc finger and BTB domain-containing protein 27 (ZBTB27)|B cell CLL/lymphoma 6|LAZ3|BCL-5|BCL6A|Protein LAZ-3|Zinc finger protein 51|BCL6_HUMAN|zinc finger transcription factor BCL6S|cys-his2 zinc finger transcription factor|Lymphoma associated zinc finger gene on chromosome 3|ZNF51|cys his2 zinc finger transcription factor|LAZ 3|Zinc finger and BTB domain-containing protein 27|B-cell lymphoma 6 protein|ZNF 51|BCL-6|ZBTB27|LAZ 3 protein|Zinc Finger Protein 51 (ZNF51)|B-cell lymphoma 5 protein
- B cell lymphoma 6 protein|ZBTB 27|Bcl 6|BCL 5|BCL6|BCL5|Zinc finger and BTB domain containing protein 27|Lymphoma Associated Zinc Finger Gene On Chromosome 3 (LAZ3)|Zinc finger and BTB domain-containing protein 27 (ZBTB27)|B cell CLL/lymphoma 6|LAZ3|BCL-5|BCL6A|Protein LAZ-3|Zinc finger protein 51|BCL6_HUMAN|zinc finger transcription factor BCL6S|cys-his2 zinc finger transcription factor|Lymphoma associated zinc finger gene on chromosome 3|ZNF51|cys his2 zinc finger transcription factor|LAZ 3|Zinc finger and BTB domain-containing protein 27|B-cell lymphoma 6 protein|ZNF 51|BCL-6|ZBTB27|LAZ 3 protein|Zinc Finger Protein 51 (ZNF51)|B-cell lymphoma 5 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41182
- P41182
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human BCL6
- Synthetic peptide of Human BCL6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together