Anti-ATP1A3 Rabbit Polyclonal Antibody (Biotin)
New Product
Type:
Antigen: ATP1A3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77712-200
- 77712-201
- Antigen Symbol:
- ATP1A3
- ATP1A3
- Antigen Synonyms:
- Alpha(III)|Na+/K+ ATPase 3|Atp1a3|DYT12|AHC2|MGC13276|Sodium pump subunit alpha-3|Sodium/potassium-transporting ATPase subunit alpha-3|Sodium/potassium transporting ATPase alpha 3 chain|Sodium pump 3|Sodium potassium ATPase alpha 3 polypeptide|DYT 12|ATPase Na+/K+ transporting alpha 3 polypeptide|Na(+)/K(+) ATPase alpha(III) subunit|RDP|Na+/K+ ATPase alpha 3 subunit|Na(+)/K(+) ATPase alpha-3 subunit|AT1A3_HUMAN
- Alpha(III)|Na+/K+ ATPase 3|Atp1a3|DYT12|AHC2|MGC13276|Sodium pump subunit alpha-3|Sodium/potassium-transporting ATPase subunit alpha-3|Sodium/potassium transporting ATPase alpha 3 chain|Sodium pump 3|Sodium potassium ATPase alpha 3 polypeptide|DYT 12|ATPase Na+/K+ transporting alpha 3 polypeptide|Na(+)/K(+) ATPase alpha(III) subunit|RDP|Na+/K+ ATPase alpha 3 subunit|Na(+)/K(+) ATPase alpha-3 subunit|AT1A3_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13637
- P13637
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sodium/potassium-transporting ATPase subunit alpha-3 protein (143-278AA)
- Recombinant Human Sodium/potassium-transporting ATPase subunit alpha-3 protein (143-278AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together